accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 4593-38-8

IUPAC Name: 5-methyl-2,3-dihydro-1H-inden-1-one
Synonyms: 5-methyl-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: KBHCTNGQJOEDDC-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=C1)C(=O)CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results