accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO

CAS RN 4593-90-2

IUPAC Name: 3-phenylbutanoic acid
Synonyms: β-phenylbutyric acid β-methylbenzenepropanoic acid 3-phenyl-N-butyric acid β-methylbenzenepropanoate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: ZZEWMYILWXCRHZ-UHFFFAOYNA-N
SMILES: CC(CC(O)=O)C1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results