accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 4594-78-9

IUPAC Name: 3-(2-oxocyclohexyl)propanenitrile
Synonyms: 3-(2-oxocyclohexyl)propanenitrile
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: SPTVCXKSSRBTMN-UHFFFAOYNA-N
SMILES: O=C1CCCCC1CCC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)