accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4605-14-5

IUPAC Name: (3-aminopropyl)({3-[(3-aminopropyl)amino]propyl})amine
Synonyms: norspermine|3,3,3-tetramine
Molecular Weight (g/mol): 188.32
Molecular Formula: C9H24N4
InChi Key: ZAXCZCOUDLENMH-UHFFFAOYSA-N
SMILES: NCCCNCCCNCCCN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results