accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 461-81-4

IUPAC Name: 1-chloro-4-(trifluoromethoxy)benzene
Synonyms: 1-chloro-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 196.55
Molecular Formula: C7H4ClF3O
InChi Key: SELFZOLQRDPBKC-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results