accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSFFF

CAS RN 461-84-7

IUPAC Name: 4-[(trifluoromethyl)sulfanyl]phenol
Synonyms: 4-[(trifluoromethyl)sulfanyl]phenol
Molecular Weight (g/mol): 194.17
Molecular Formula: C7H5F3OS
InChi Key: YYCPTWHVKSATQK-UHFFFAOYSA-N
SMILES: OC1=CC=C(SC(F)(F)F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)