accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4612-28-6

IUPAC Name: [3-(dihydroxyboranyl)phenyl]boronic acid
Synonyms: 3-(dihydroxyboranyl)phenylboronic acid
Molecular Weight (g/mol): 165.75
Molecular Formula: C6H8B2O4
InChi Key: UXPAASVRXBULRG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results