accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)O(S)(R)HOO(R)HOHO(S)OH(R)OH(S)OH(R)HO

CAS RN 4618-18-2

IUPAC Name: (2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: lactulose portalac cephulac duphalac generlac bifiteral isolactose constilac enulose lactulose, ~98%
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: JCQLYHFGKNRPGE-FCVZTGTOSA-N
SMILES: OC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (6)
  • (1)
Physical Form 
  • (5)
  • (6)
Quantity 
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
  • (3)
Grade 
  • (1)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (4)
  • (6)
  • (1)
  • (5)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (5)