CAS RN 46181-30-0
IUPAC Name:
2-methyl-6-phenylpyridine
Synonyms:
2-methyl-6-phenylpyridine
Molecular Weight (g/mol):
169.23
Molecular Formula:
C12H11N
InChi Key:
GREMYQDDZRJQEG-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Methyl-6-phenylpyridine 98.0+%, TCI America™
CAS: 46181-30-0 Molecular Formula: C12H11N Molecular Weight (g/mol): 169.23 MDL Number: MFCD00796537 InChI Key: GREMYQDDZRJQEG-UHFFFAOYSA-N PubChem CID: 2762885 IUPAC Name: 2-methyl-6-phenylpyridine SMILES: CC1=CC=CC(=N1)C1=CC=CC=C1