accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 4619-20-9

IUPAC Name: 4-(4-methylphenyl)-4-oxobutanoic acid
Synonyms: 4-(4-methylphenyl)-4-oxobutanoic acid
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: OEEUWZITKKSXAZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)CCC(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)