accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 46196-54-7

IUPAC Name: 3-(2-methylphenoxy)pyrrolidine
Synonyms: 3-(2-methylphenoxy)pyrrolidine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: MIVMFYNEGPHLSB-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1OC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results