accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 462-34-0

IUPAC Name: oxolane; trifluoroborane
Synonyms: THF; boron trifluoride
Molecular Weight (g/mol): 139.91
Molecular Formula: C4H8BF3O
InChi Key: CHNLPLHJUPMEOI-UHFFFAOYSA-N
SMILES: FB(F)F.C1CCOC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)