accessibility menu, dialog, popup

UserName

Viewing profile as user:
FF

CAS RN 462-39-5

IUPAC Name: 1,3-difluoropropane
Synonyms: 1,3-difluoropropane
Molecular Weight (g/mol): 80.08
Molecular Formula: C3H6F2
InChi Key: OOLOYCGJRJFTPM-UHFFFAOYSA-N
SMILES: FCCCF
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)