accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBr

CAS RN 462-72-6

IUPAC Name: 1-bromo-4-fluorobutane
Synonyms: 1-bromo-4-fluorobutane
Molecular Weight (g/mol): 155.01
Molecular Formula: C4H8BrF
InChi Key: WMCUHRDQSHQNRW-UHFFFAOYSA-N
SMILES: FCCCCBr
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)