accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4628-94-8

IUPAC Name: dipotassium (5-sulfanidyl-1,3,4-thiadiazol-2-yl)sulfanide
Synonyms: dipotassium (5-sulfanidyl-1,3,4-thiadiazol-2-yl)sulfanide
Molecular Weight (g/mol): 226.41
Molecular Formula: C2K2N2S3
InChi Key: GPWLFGDMYSVEGN-UHFFFAOYSA-L
SMILES: [K+].[K+].[S-]C1=NN=C([S-])S1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results