accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO(E)(E)(E)

CAS RN 463-40-1

IUPAC Name: (9E,12E,15E)-octadeca-9,12,15-trienoic acid
Synonyms: elaidolinoleic acid
Molecular Weight (g/mol): 278.44
Molecular Formula: C18H30O2
InChi Key: DTOSIQBPPRVQHS-IUQGRGSQSA-N
SMILES: CC\C=C\C\C=C\C\C=C\CCCCCCCC(O)=O
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)