CAS RN 463-71-8
IUPAC Name:
chloromethanecarbothioyl chloride
Synonyms:
thiophosgene cscl2 carbonothioic dichloride thiocarbonic dichloride
Molecular Weight (g/mol):
114.97
Molecular Formula:
CCl2S
InChi Key:
ZWZVWGITAAIFPS-UHFFFAOYSA-N
SMILES:
ClC(Cl)=S
1
–
2
of
2
results
1
Promotions Available
Thiophosgene, 85%
Thiophosgene, 85%
CAS: 463-71-8 MDL Number: MFCD00004918 InChI Key: ZWZVWGITAAIFPS-UHFFFAOYSA-N Synonym: thiophosgene,carbonothioic dichloride,carbon chlorosulfide,thiocarbonyl chloride,thiocarbonic dichloride,dichlorothiocarbonyl,thiofosgen,phosgene, thio,thiokarbonylchlorid,carbonyl chloride, thio PubChem CID: 10040 ChEBI: CHEBI:29366 IUPAC Name: thiocarbonyl dichloride SMILES: C(=S)(Cl)Cl
2
Sigma Aldrich Thiophosgene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.