accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 4630-82-4

IUPAC Name: methyl cyclohexanecarboxylate
Synonyms: cyclohexanecarboxylic acid, methyl ester
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: ZQWPRMPSCMSAJU-UHFFFAOYSA-N
SMILES: COC(=O)C1CCCCC1
Boiling Point 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)