accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 4634-13-3

IUPAC Name: methyl 6-phenylpyridine-3-carboxylate
Synonyms: methyl 6-phenylpyridine-3-carboxylate
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: DXHZSBAEWGPUKI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results