accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4635-60-3

IUPAC Name: 4-chloro-N,N-bis(propan-2-yl)butanamide
Synonyms: 4-chloro-N,N-diisopropylbutanamide
Molecular Weight (g/mol): 205.73
Molecular Formula: C10H20ClNO
InChi Key: HOSYQMBAZLKTJR-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results