accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4638-92-0

IUPAC Name: (1S,3R)-2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylic acid
Synonyms: (-)-cis-chrysanthemic acid (-)-cis-chrysanthemumsaeure
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: XLOPRKKSAJMMEW-HTQZYQBOSA-N
SMILES: CC(C)=C[C@@H]1[C@H](C(O)=O)C1(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results