CAS RN 464-45-9
(-)-Borneol, 97+%
CAS: 464-45-9 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 MDL Number: MFCD00003759,MFCD00066426,MFCD00066427 InChI Key: DTGKSKDOIYIVQL-NQMVMOMDSA-N Synonym: --borneol,borneol,l-borneol,1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol,linderol,1s-endo---borneol,borneo camphor,bornyl alcohol,sumatra camphor,dl-borneol PubChem CID: 1201518 ChEBI: CHEBI:15394 SMILES: CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
L-(-)-Borneol, 98.0%, MP Biomedicals™
CAS: 464-45-9 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 MDL Number: MFCD00003759,MFCD00066426,MFCD00066427 InChI Key: DTGKSKDOIYIVQL-NQMVMOMDSA-N Synonym: --borneol,borneol,l-borneol,1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol,linderol,1s-endo---borneol,borneo camphor,bornyl alcohol,sumatra camphor,dl-borneol PubChem CID: 1201518 ChEBI: CHEBI:15394 IUPAC Name: (1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol SMILES: CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
(-)-Borneol, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(-)-Borneol is an enantiomer. It is a bicyclic monoterpene compound used gengrally for analgesia and anaesthesia. It is considered as positive modulators of GABA receptors.
phyproof™ Borneol, ≥95% (GC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
L(-)-Borneol, 97%
CAS: 464-45-9 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 MDL Number: MFCD00003759,MFCD00066426,MFCD00066427 InChI Key: DTGKSKDOIYIVQL-NQMVMOMDSA-N Synonym: --borneol,borneol,l-borneol,1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol,linderol,1s-endo---borneol,borneo camphor,bornyl alcohol,sumatra camphor,dl-borneol PubChem CID: 1201518 ChEBI: CHEBI:15394 IUPAC Name: (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol SMILES: CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
(-)-Borneol 95.0+%, TCI America™
CAS: 464-45-9 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.25 MDL Number: MFCD00003759,MFCD00066426,MFCD00066427 InChI Key: DTGKSKDOIYIVQL-NQMVMOMDSA-N Synonym: --borneol,borneol,l-borneol,1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol,linderol,1s-endo---borneol,borneo camphor,bornyl alcohol,sumatra camphor,dl-borneol PubChem CID: 1201518 ChEBI: CHEBI:15394 IUPAC Name: (1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol SMILES: CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
Sigma Aldrich (-)-Borneol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.