accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3(E)

CAS RN 4643-27-0

IUPAC Name: (2E)-oct-2-en-4-one
Synonyms: (2E)-oct-2-en-4-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: FMDLEUPBHMCPQV-GQCTYLIASA-N
SMILES: CCCCC(=O)\C=C\C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results