accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4643-36-1

H3CNHHCl

CAS RN 4643-36-1

IUPAC Name: N-methylnaphthalen-1-amine hydrochloride
Synonyms: 1-naphthalenamine, N-methyl- hydrochloride
Molecular Weight (g/mol): 193.67
Molecular Formula: C11H12ClN
InChi Key: ALZGTEASAJRPTL-UHFFFAOYSA-N
SMILES: Cl.CNC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)