accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4651-81-4

IUPAC Name: methyl 2-aminothiophene-3-carboxylate
Synonyms: methyl 2-aminothiophene-3-carboxylate
Molecular Weight (g/mol): 157.19
Molecular Formula: C6H7NO2S
InChi Key: DGGJQLCAYQCPDD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results