accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSN

CAS RN 4651-91-6

IUPAC Name: 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Synonyms: 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Molecular Weight (g/mol): 178.25
Molecular Formula: C9H10N2S
InChi Key: ADHVMGAFAKSNOM-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C2=C(CCCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results