accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4653-08-1

IUPAC Name: 4-oxo-4-(thiophen-2-yl)butanoic acid
Synonyms: 4-oxo-4-(thiophen-2-yl)butanoic acid
Molecular Weight (g/mol): 184.21
Molecular Formula: C8H8O3S
InChi Key: ULJMYWHLMLRYSO-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results