accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH2

CAS RN 4655-34-9

IUPAC Name: propan-2-yl 2-methylprop-2-enoate
Synonyms: isopropyl methacrylate N-propyl methacrylate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: BOQSSGDQNWEFSX-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results