accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 465514-33-4

IUPAC Name: [2-(morpholin-4-yl)phenyl]methanol
Synonyms: [2-(morpholin-4-yl)phenyl]methanol
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: MYGVYNRBQSAMIF-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results