accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHONNFFF

CAS RN 465514-39-0

IUPAC Name: 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
Synonyms: 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
Molecular Weight (g/mol): 275.23
Molecular Formula: C11H12F3N3O2
InChi Key: QYAIISDWPUEPHG-UHFFFAOYSA-N
SMILES: OC(=O)C1CCN(CC1)C1=NC=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results