CAS RN 466-99-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Hydromorphone Solution, 1 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
A certified reference standard suitable for use as a starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research.