CAS RN 4669-59-4
CAS RN 4669-59-4
IUPAC Name:
[(diethylamino)dimethylsilyl]diethylamine
Synonyms:
[(diethylamino)dimethylsilyl]diethylamine
Molecular Weight (g/mol):
202.42
Molecular Formula:
C10H26N2Si
InChi Key:
XIFOKLGEKUNZTI-UHFFFAOYSA-N
SMILES:
CCN(CC)[Si](C)(C)N(CC)CC
1
–
1
of
1
results
1
Bis(diethylamino)dimethylsilane 98.0+%, TCI America™
CAS: 4669-59-4 Molecular Formula: C10H26N2Si Molecular Weight (g/mol): 202.417 MDL Number: MFCD00039879 InChI Key: XIFOKLGEKUNZTI-UHFFFAOYSA-N Synonym: N,N,N′,N′-Tetraethyl-1,1-dimethylsilanediamine PubChem CID: 78399 IUPAC Name: N-[diethylamino(dimethyl)silyl]-N-ethylethanamine SMILES: CCN(CC)[Si](C)(C)N(CC)CC