accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(R)OO(S)(S)(R)(S)(S)OH(S)H3C(S)O(S)H3C(R)(S)H3C

CAS RN 467-55-0

IUPAC Name: (1'R,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'S,16'S,18'S)-16'-hydroxy-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-10'-one
Synonyms: hecogenin hocogenin
Molecular Weight (g/mol): 430.63
Molecular Formula: C27H42O4
InChi Key: QOLRLLFJMZLYQJ-LOBDNJQFSA-N
SMILES: C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC[C@H]5C[C@@H](O)CC[C@]5(C)[C@H]4CC(=O)[C@]23C)O[C@]11CC[C@@H](C)CO1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)