accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHO

CAS RN 467-69-6

IUPAC Name: 9-hydroxy-9H-fluorene-9-carboxylic acid
Synonyms: flurenol
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: GXAMYUGOODKVRM-UHFFFAOYSA-N
SMILES: OC(=O)C1(O)C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results