accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3C

CAS RN 4670-10-4

IUPAC Name: 2-(3,5-dimethoxyphenyl)acetate
Synonyms: 2-(3,5-dimethoxyphenyl)acetate
Molecular Weight (g/mol): 195.2
Molecular Formula: C10H11O4
InChi Key: FFPAFDDLAGTGPQ-UHFFFAOYSA-M
SMILES: COC1=CC(OC)=CC(CC([O-])=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results