accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4672-30-4

IUPAC Name: (2-phenylethyl)phosphonic acid
Synonyms: 2-phenylethylphosphonic acid
Molecular Weight (g/mol): 186.15
Molecular Formula: C8H11O3P
InChi Key: BFDYSJCMAFSRDH-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results