accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4674-50-4

IUPAC Name: 4,4a-dimethyl-6-(prop-1-en-2-yl)-2,3,4,4a,5,6,7,8-octahydronaphthalen-2-one
Synonyms: nootkatone
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: WTOYNNBCKUYIKC-UHFFFAOYNA-N
SMILES: CC1CC(=O)C=C2CCC(CC12C)C(C)=C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)