accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 467435-07-0

BrClFFF

CAS RN 467435-07-0

IUPAC Name: 4-bromo-2-chloro-1-(trifluoromethyl)benzene
Synonyms: 4-bromo-2-chloro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: FJANWAMBKVZTRG-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results