accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 468-44-0

IUPAC Name: (1R,2R,5R,8R,9S,10R,12S)-12-hydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadecane-9-carboxylic acid
Synonyms: (1R,2R,5R,8R,9S,10R,12S)-12-hydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadecane-9-carboxylic acid
Molecular Weight (g/mol): 332.4
Molecular Formula: C19H24O5
InChi Key: RSQSQJNRHICNNH-WTVOOMFRSA-N
SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45C[C@@H](CC[C@H]4[C@@]3(CC[C@@H]1O)OC2=O)C(=C)C5

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results