accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4685-47-6

IUPAC Name: 4-methoxy-1,2-dimethylbenzene
Synonyms: 4-methoxy-1,2-dimethyl-benzene
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: LVUBSVWMOWKPDJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)