accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 4694-80-8

IUPAC Name: 2-propyl-4,5-dihydro-1,3-oxazole
Synonyms: 2-propyl-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: GXCJLVVUIVSLOQ-UHFFFAOYSA-N
SMILES: CCCC1=NCCO1
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results