accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 469865-01-8

IUPAC Name: 10,10'-dimethyl-[9,9'-biacridine]-10,10'-diium bis(4-(methoxycarbonyl)benzoate)
Synonyms: 10,10'-dimethyl-[9,9'-biacridine]-10,10'-diium bis(4-(methoxycarbonyl)benzoate)
Molecular Weight (g/mol): 744.8
Molecular Formula: C46H36N2O8
InChi Key: BPSROIZFFFEAQI-UHFFFAOYSA-L
SMILES: COC(=O)C1=CC=C(C=C1)C([O-])=O.COC(=O)C1=CC=C(C=C1)C([O-])=O.C[N+]1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results