accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3O

CAS RN 470-67-7

IUPAC Name: 1-methyl-4-(propan-2-yl)-7-oxabicyclo[2.2.1]heptane
Synonyms: 1,4-cineol isocineole
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: RFFOTVCVTJUTAD-UHFFFAOYSA-N
SMILES: CC(C)C12CCC(C)(CC1)O2
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)