accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 4702-34-5

IUPAC Name: 3,4-dihydro-1H-2-benzopyran-1-one
Synonyms: 3,4-dihydro-2-benzopyran-1-one
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: XVTAQSGZOGYIEY-UHFFFAOYSA-N
SMILES: O=C1OCCC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results