accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4704-77-2

IUPAC Name: 3-bromopropane-1,2-diol
Synonyms: 3-bromo-1,2-propanediol C3H7BrO2 bromodeoxyglycerol α-bromohydrin monobromoglycerol glycerol bromohydrin 3-bromodeoxyglycerol
Molecular Weight (g/mol): 154.99
Molecular Formula: C3H7BrO2
InChi Key: SIBFQOUHOCRXDL-UHFFFAOYNA-N
SMILES: OCC(O)CBr
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)