accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4705-34-4

IUPAC Name: 1-methoxy-4-[(1E)-2-(4-methoxyphenyl)ethenyl]benzene
Synonyms: 1-methoxy-4-[(1E)-2-(4-methoxyphenyl)ethenyl]benzene
Molecular Weight (g/mol): 240.3
Molecular Formula: C16H16O2
InChi Key: CAWFCZIEFIQKRV-ONEGZZNKSA-N
SMILES: COC1=CC=C(\C=C\C2=CC=C(OC)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results