accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4707-95-3

CH3CH3H3CPOClCl

CAS RN 4707-95-3

IUPAC Name: tert-butylphosphonoyl dichloride
Synonyms: tert-butylphosphonoyl dichloride
Molecular Weight (g/mol): 174.99
Molecular Formula: C4H9Cl2OP
InChi Key: OQUGNSMOWJIYMO-UHFFFAOYSA-N
SMILES: CC(C)(C)P(Cl)(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results