accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(E/Z)

CAS RN 4714-21-0

IUPAC Name: 1-methyl-4-(2-phenylethenyl)benzene
Synonyms: 1-methyl-4-(2-phenylethenyl)benzene
Molecular Weight (g/mol): 194.28
Molecular Formula: C15H14
InChi Key: MDRVHDXASYPUCB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results