accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4714-62-9

IUPAC Name: 4-(methylamino)benzonitrile
Synonyms: 4-(methylamino)benzonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: JEDXDYXIRZOEKG-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results