CAS RN 47162-00-5
IUPAC Name:
1-{[1,1'-biphenyl]-4-yl}octan-1-one
Synonyms:
1-{[1,1'-biphenyl]-4-yl}octan-1-one
Molecular Weight (g/mol):
280.41
Molecular Formula:
C20H24O
InChi Key:
JSNGEMRLSPQOOF-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-n-Octanoylbiphenyl 97.0+%, TCI America™
CAS: 47162-00-5 Molecular Formula: C20H24O Molecular Weight (g/mol): 280.411 MDL Number: MFCD00143358 InChI Key: JSNGEMRLSPQOOF-UHFFFAOYSA-N PubChem CID: 607296 IUPAC Name: 1-(4-phenylphenyl)octan-1-one SMILES: CCCCCCCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2